Learn More
R&L Enterprises™ Cork Borer Sharpner
Product Code. 12883952 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
1Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12883952 1Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12883952
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 01-04-2027
Add to basket
Add to basket
Keeps cork borers sharp and efficient to use. R&L Enterprises™ Cork Borer Sharpeners are for cork borers from 4 to 27mm.
  • With metal handle
  • Metal cone with hinged cutter blade set in slot
Specifications
Product Type Cork Borer Sharpener
For Use With (Application) For 4 to 27mm dia. Borers
Quantity 1Each