missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated PARD3 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

1500.75 NOK - 4060.17 NOK

Specifications

Antigen PARD3
Clone 1D2B2
Concentration 1000 μg/mL
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233170
View Documents
Proteintech
CL488-66968-20UL
20 μL
ONLINE EXCLUSIVE PRICE
1651.35 NOK
1500.75 NOK / 20µL
Save 150.60 NOK
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30236116
View Documents
Proteintech
CL488-66968-100UL
100 μL
ONLINE EXCLUSIVE PRICE
5307.92 NOK
4060.17 NOK / 100µL
Save 1247.75 NOK
23% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PARD3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human antigen. The PARD3 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

PARD3
1000 μg/mL
Monoclonal
Liquid
RUO
56288
Recombinant Protein
Primary
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
1D2B2
Immunocytochemistry/Immunofluorescence
CoraLite Plus 488
Mouse
Human
ASIP; Baz; Bazooka; FLJ21015; PAR3; PAR3alpha; PARD3A; SE2-5L16; SE2-5LT1; SE2-5T2
IgG1
Protein G purification
Conjugated Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.