Learn More
CoraLite™ Plus 488-conjugated PARD3 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66968-20UL
This item is not returnable.
View return policy
Description
The PARD3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human antigen. The PARD3 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Specifications
| PARD3 | |
| Monoclonal | |
| 1000 μg/mL | |
| ASIP; Baz; Bazooka; FLJ21015; PAR3; PAR3alpha; PARD3A; SE2-5L16; SE2-5LT1; SE2-5T2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 170 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 1D2B2 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 56288 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG1 |