missing translation for 'onlineSavingsMsg'
Learn More

COPZ1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20440-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16899974

  • 1415.13 NOK / 20µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

COPZ1
Polyclonal
Unconjugated
COPZ1
CGI 120, Coatomer subunit zeta 1, COPZ, COPZ1, HSPC181, Zeta 1 coat protein, Zeta 1 COP, zeta1 COP
Rabbit
Antigen Affinity Chromatography
RUO
22818, 315345, 56447
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P61923, P61924
Copz1
COPZ1 Fusion Protein Ag14234
20 μL
Primary
Rat, Mouse, Human, Non-human Primate
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers