missing translation for 'onlineSavingsMsg'
Learn More

COPZ1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen COPZ1
Concentration 0.15 mg/mL
Applications Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899974
View Documents
Proteintech
20440-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16889974
View Documents
Proteintech
20440-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a subunit of the cytoplasmic coatamer protein complex, which is involved in autophagy and intracellular protein trafficking. The coatomer protein complex is comprised of seven subunits and functions as the coat protein of coat protein complex (COP)I-vesicles. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

COPZ1
Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Rat, Mouse, Human, Non-human Primate
P61923, P61924
22818, 315345, 56447
COPZ1 Fusion Protein Ag14234
Primary
-20°C
COPZ1
0.15 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CGI 120, Coatomer subunit zeta 1, COPZ, COPZ1, HSPC181, Zeta 1 coat protein, Zeta 1 COP, zeta1 COP
Copz1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.