Learn More
Eppendorf™ Centrifuge 5920 R
BIOPHARMAGREENER_CHOICE
Product Code. 16000552 Shop All Eppendorf Products
Change view
Click to view available options
Includes:
Rotor, Plate/Tube Bucket, Plate Carrier
Rotor, Plate/Tube Buckets, Adapters for 15/50 mL Conical Tubes
Rotor, Plate/Tube Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.)
Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes
Rotor, Universal Buckets, Adapters for 5/15/25/50 mL Conical Tubes and Plates
Rotor, Universal Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.)
Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes
Without Rotor
Model:
Centrifuge 5920 R
Centrifuge 5920 R (EU-IVD)
Rotor:
Not Included
S-4 xUniversal-Large
S-4x1000
S-4x750
Electrical Requirements:
230 V, 50/60 Hz (EU)
Unit Size:
1 set
Each
Product Code. Includes Model Rotor Electrical Requirements unitSize
16000552 Rotor, Universal Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.) Centrifuge 5920 R (EU-IVD) S-4 xUniversal-Large 230 V, 50/60 Hz (EU) Each
16574144 Rotor, Universal Buckets, Adapters for 5/15/25/50 mL Conical Tubes and Plates Centrifuge 5920 R (EU-IVD) S-4 xUniversal-Large 230 V, 50/60 Hz (EU) 1 set
15743315 Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R (EU-IVD) S-4x750 230 V, 50/60 Hz (EU) Each
15694614 Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R (EU-IVD) S-4x1000 230 V, 50/60 Hz (EU) Each
15638610 Rotor, Plate/Tube Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R (EU-IVD) S-4x1000 230 V, 50/60 Hz (EU) Each
15763315 Rotor, Plate/Tube Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.) Centrifuge 5920 R (EU-IVD) S-4x1000 230 V, 50/60 Hz (EU) Each
15713315 Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R (EU-IVD) S-4x1000 230 V, 50/60 Hz (EU) Each
15885123 Without Rotor Centrifuge 5920 R Not Included 230 V, 50/60 Hz (EU) Each
16564144 Rotor, Universal Buckets, Adapters for 5/15/25/50 mL Conical Tubes and Plates Centrifuge 5920 R S-4 xUniversal-Large 230 V, 50/60 Hz (EU) 1 set
15815143 Rotor, Universal Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.) Centrifuge 5920 R S-4 xUniversal-Large 230 V, 50/60 Hz (EU) 1 set
15865133 Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R S-4x750 230 V, 50/60 Hz (EU) 1 set
15805133 Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R S-4x1000 230 V, 50/60 Hz (EU) 1 set
16652842 Rotor, Plate/Tube Bucket, Plate Carrier Centrifuge 5920 R S-4x1000 230 V, 50/60 Hz (EU) Each
15845133 Rotor, Plate/Tube Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R S-4x1000 230 V, 50/60 Hz (EU) 1 set
15825133 Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes Centrifuge 5920 R S-4x1000 230 V, 50/60 Hz (EU) 1 set
15 options
Product Code. 16000552
Brand: Eppendorf™ 5948000816
The refrigerated Eppendorf Centrifuge 5920 R offers high-throughput performance and advanced temperature management in a very compact design. With a capacity of up to 4 x 1,000 mL / 52 x 50 mL conical tubes, it can spin both plates and tubes in the same bucket for real flexibility.
cooled, with rotor S-4xUniversal-Large, incl. Universal beakers and adapters for conical tubes 5 mL / 15 mL / 25 mL / 50 mL and plates, 230 V / 50 - 60 Hz
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
  • Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
  • High centrifugation speed of up to 21,194 x g (13,700 rpm)
  • Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking
  • Centrifuge lid with soft-touch lid closure
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Controller Type Keypad
Max. RCF 21,194 x g
Max. Speed 13,000 rpm
Refrigerated Yes
Display Large, Brightly Lit LCD
Includes Rotor, Universal Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.)
Maximum Speed Range 100 to 13,700 rpm
Model Centrifuge 5920 R (EU-IVD)
Noise Level <55 dB(A)
Rotor S-4 xUniversal-Large
Rotor Included Yes
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
Standards EN 61010-1, EN 61010-2-020, EN 61326-1, EN 55011, CFR 47 FCC part 15 class A, EN 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-3, EN ISO 18113-1, EN ISO 15223-1, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
Timer 10 sec. to 99 hr. 59 min., Continuous Run Function, Short-spin
Type Benchtop Centrifuge
Length (Metric) Exterior 71 cm
Width (English) Exterior 29.2 in.
Width (Metric) Exterior 74 cm
Height (English) Exterior 15.9 in.
Height (Metric) Exterior 40 cm
Net Weight 139 kg
Electrical Requirements 230 V, 50/60 Hz (EU)
Voltage 230 V
Wattage 1650 W
Frequency 50/60 Hz
Show More Show Less
The Centrifuge 5920 R is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.