Learn More
Eppendorf™ Centrifuge 5920 R Product Code.: 15694614
Eppendorf™ Centrifuge 5920 R
GREENER_CHOICE
Product Code. 15694614 Shop All Eppendorf Products
Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes, Centrifuge 5920 R (EU-IVD), S-4x1000, 230 V, 50/60 Hz (EU), Each
Click to view available options
Includes:
Rotor, Plate/Tube Bucket, Plate Carrier
Rotor, Plate/Tube Buckets, Adapters for 15/50 mL Conical Tubes
Rotor, Plate/Tube Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.)
Rotor, Round Buckets, Adapters for 15/50 mL Conical Tubes
Rotor, Universal Buckets, Adapters for 5/15/25/50 mL Conical Tubes and Plates
Rotor, Universal Buckets, Adapters for Round-bottom Tubes (13/16 mm dia.)
Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes
Without Rotor
Model:
Centrifuge 5920 R
Centrifuge 5920 R (EU-IVD)
Rotor:
Not Included
S-4 xUniversal-Large
S-4x1000
S-4x750
Electrical Requirements:
230 V, 50/60 Hz (EU)
Unit Size:
1 set
Each
Product Code. 15694614
Brand: Eppendorf™ 5948000212
The refrigerated Eppendorf Centrifuge 5920 R offers high-throughput performance and advanced temperature management in a very compact design. With a capacity of up to 4 x 1,000 mL / 52 x 50 mL conical tubes, it can spin both plates and tubes in the same bucket for real flexibility.
cooled, with rotor S-4xUniversal-Large, incl. Universal beakers and adapters for conical tubes 5 mL / 15 mL / 25 mL / 50 mL and plates, 230 V / 50 - 60 Hz
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
  • Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
  • High centrifugation speed of up to 21,194 x g (13,700 rpm)
  • Aerosol-tight Eppendorf QuickLock™ caps and lids allow fast and ergonomic lid locking
  • Centrifuge lid with soft-touch lid closure
  • Quiet operation to improve your work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
    • Free-of a substance that causes environmental harm
    • Recycled Content
    • Non-Toxic
    • Recyclable
    • Ozone-Safe or Safe for the Ozone Layer
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers
Controller Type Keypad
Description Centrifuge 5920 R
Capacity 4 x 1000 mL
Max. RCF 21,194 x g
Max. Speed 13,500 rpm
Refrigerated Yes
Display Large, Brightly Lit LCD
Includes Rotor, high-capacity Buckets, Adapters for 15/50 mL Conical Tubes
Model Centrifuge 5920 R (EU-IVD)
Noise Level <60 dB(A)
Rotor S-4x1000
Rotor Included Yes
Run Time 10 sec. to 99 hr. 59 min., Continuous Run Function, Short-spin
Safety Features Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids and aerosol-tight caps, Rotor lifecycle counter, automatic imbalance recognition
Standards EN 61010-1, EN 61010-2-020, EN 61326-1, EN 55011, CFR 47 FCC part 15 class A, EN 14971, EN 61010-2-101, EN 61326-2-6, EN 62366, EN ISO 18113-3, EN ISO 18113-1, EN ISO 15223-1, EN 378-1, EN 378-2, EN 50581 (refer to manual for more details)
Temperature Range -11°C to +40°C
Timer 10 sec. to 99 hr. 59 min., Continuous Run Function, Short-spin
Type Benchtop Centrifuge
Warranty 3 Years
Dimensions (HWD) 70 x 61 x 35 cm
Width (English) Exterior 29.2 in.
Width (Metric) Exterior 74 cm
Height (English) Exterior 15.9 in.
Height (Metric) Exterior 40 cm
Net Weight 99 kg
Electrical Requirements 230 V, 50/60 Hz (EU)
Voltage 230 V
Wattage 1650 W
Show More Show Less
The Centrifuge 5920 R is designed for separating liquid substance mixtures with different densities, in particular, for processing and analyzing samples from the human body in in-vitro diagnostic applications to ensure that the in-vitro diagnostic device can be used according to its intended purpose. This centrifuge including components is an in-vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. Eppendorf centrifuges are exclusively intended for indoor use by trained specialists.
Product Content Correction
Product Title
Eppendorf™ Centrifuge 5920 R >