Learn More
Invitrogen™ Control BACULOSOMES™ Plus Reagent
Product Code. 11589236
Click to view available options
Quantity:
5 mg
Unit Size:
Each
Product Code. 11589236
Brand: Invitrogen™ P2315
Control BACULOSOMES™ Plus Reagent
Control BACULOSOMES™ Plus Reagent is a microsomal preparation from insect cells infected with wild-type baculovirus. This product is suitable as a negative control for assays carried out using our CYP450 BACULOSOMES™ Plus Reagents.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY
Specifications
Product Line BACULOSOMES™
Content And Storage Store at -80°C
Product Type Bacterial Microsome
Quantity 5 mg
Format Liquid
Shipping Condition Dry Ice
Research Category Toxicology & Drug Metabolism