missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 45/GJC1 Antibody (5B9.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29754-0.025MG
This item is not returnable.
View return policy
Description
Connexin 45/GJC1 Monoclonal specifically detects Connexin 45/GJC1 in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Connexin 45/GJC1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| P36383 | |
| GJC1 | |
| The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-377 of human connexin 45. | |
| 0.025 mg | |
| Primary | |
| Recognizes Human Connexin 45, also confirmed to work on rat C6 cells. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 5B9.2 | |
| Unconjugated | |
| 20mM Phosphate (pH 7.6) and 0.25M NaCl with 0.1% Sodium Azide | |
| connexin 45, Connexin-45, CX45, DKFZp686P0738, Gap junction alpha-7 protein, gap junction gamma-1 protein, gap junction protein, alpha 7, 45kDa, gap junction protein, gamma 1, 45kDa, GJA7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 10052 | |
| Rat, Human, Rabbit | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction