missing translation for 'onlineSavingsMsg'
Learn More

Connexin 45/GJC1 Antibody (5B9.2) - BSA Free, Novus Biologicals™

Product Code. p-7200850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
Product Code. Quantity unitSize
18124843 0.025 mg 25µg
18144335 0.1 mg 0.1mg
2 options
This item is not returnable. View return policy

Product Code. 18124843

Brand: Novus Biologicals NBP2297540.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Connexin 45/GJC1 Monoclonal specifically detects Connexin 45/GJC1 in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 45/GJC1
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5B9.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation 20mM Phosphate (pH 7.6) and 0.25M NaCl with 0.1% Sodium Azide
Gene Accession No. P36383
Gene Alias connexin 45, Connexin-45, CX45, DKFZp686P0738, Gap junction alpha-7 protein, gap junction gamma-1 protein, gap junction protein, alpha 7, 45kDa, gap junction protein, gamma 1, 45kDa, GJA7
Gene Symbols GJC1
Host Species Mouse
Immunogen The peptide sequence used for the immunogen was H-Gln-Ala-Tyr-Ser-His-Gln-Asn-Asn-Pro-His-Gly-Pro-Arg-Glu-Cys-OH conjugated to KLH, corresponding to amino acids 354-377 of human connexin 45.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10052
Test Specificity Recognizes Human Connexin 45, also confirmed to work on rat C6 cells.
Target Species Rat, Human, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.