Learn More
Eppendorf™ Conical Bottom 384 Well Twin Tec PCR Plate
Shop All Eppendorf Products
Click to view available options
Color:
Black, Clear Bottom
Black, Yellow
Blue
Green
Orange, Clear Bottom
Red
Yellow
Unit Size:
Pack of 25
Pack of 300
Specifications
Specifications
| Format | Standard |
| Array | 16 x 24 |
| Color | Blue |
| No. of Wells | 384 |
| Well Shape | Conical Bottom (V-well) |
| No. per Pack | 300 |
| Quantity | 300/Pk. |