Learn More
Fisherbrand™ Condenser High Speed
Product Code. p-7221111 Shop All Fisherbrand Products
Click to view available options
Frequency:
50 Hz
60 Hz
For Use With (Equipment):
Blue Line 5 / IQ5
Blue Line 6 / IQ6
Unit Size:
Each
Product Code. 17157804
Only null left
Add to basket
Add to basket
A combination of high speed and condenser recycling water and heat system.
  • The full Condenser Recycle Water and Heat and a half High Speed module
  • One tank provided from the High Speed module, and second tank provided from the Condenser Recycle Water and Heat
Specifications
Product Type Condenser
For Use With (Equipment) Blue Line 5 / IQ5
Frequency 60 Hz