Learn More
Fisherbrand™ Condenser High Speed
Shop All Fisherbrand Products
Click to view available options
Frequency:
50 Hz
60 Hz
For Use With (Equipment):
Blue Line 5 / IQ5
Blue Line 6 / IQ6
Unit Size:
Each
Description
- The full Condenser Recycle Water and Heat and a half High Speed module
- One tank provided from the High Speed module, and second tank provided from the Condenser Recycle Water and Heat
Specifications
Specifications
| Product Type | Condenser |
| For Use With (Equipment) | Blue Line 5 / IQ5 |
| Frequency | 50 Hz |