Learn More
Eppendorf™ Concentrator Plus Rotors
Product Code. 10391584 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
16 x 6.5/10 mL
18 x 10 mL
2 x 96 x 0.2 mL
24 x 6 mL
36 x 1.5 mL
6 x 15/50 mL
8 x 15 mL
8 x 16 mL
8 x 25 mL
Description:
Rotor A-2-VC
Rotor F-35-6-30
Rotor F-35-8-24
Rotor F-40-18-19
Rotor F-40-36-12
Rotor F-45-16-20
Rotor F-45-24-12
Rotor F-45-8-17
Rotor F-50-8-16
Rotor F-50-8-18
Unit Size:
Each
Product Code. Capacity Description unitSize
10391584 8 x 25 mL Rotor F-35-8-24 Each
10737274 18 x 10 mL Rotor F-40-18-19 Each
10337133 24 x 6 mL Rotor F-45-24-12 Each
10125802 2 x 96 x 0.2 mL Rotor A-2-VC Each
10609503 16 x 6.5/10 mL Rotor F-45-16-20 Each
10498983 36 x 1.5 mL Rotor F-40-36-12 Each
11960517 6 x 15/50 mL Rotor F-35-6-30 Each
10554244 8 x 15 mL Rotor F-45-8-17 Each
10327423 8 x 16 mL Rotor F-50-8-18 Each
10522844 8 x 15 mL Rotor F-50-8-16 Each
10 options
Product Code. 10391584
Brand: Eppendorf™ 5490039009
Designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench
  • Advanced heating technology provides best treatment for your sample which assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. Our new coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
  • The Concentrator plus is available with three configurations to meet the customers' lab specific needs:
  • Basic Concentrator for connecting to an existing vacuum pump
  • Complete system with an integrated solvent-resistant, maintenance and oil-free PTFE pump, and condensation trap
  • Complete system with vacuum port adds extra capability. It allows for using the integrated vacuum pump separately – without having to disconnect it from the system
  • Choice of 3 operation modes (vacufuge, centrifuge, and desiccator) for a variety of application needs
  • Choice of three application modes (aqueous, alcohol or high vapor pressure) to correspond with sample solvent—reduction of processing time up to 20%
  • Choice of 4 heating levels (ambient, 30C, 45°C, 60°C) allows safe and efficient concentration of various samples
  • Brake function can optionally be turned off for use with delicate samples
TRUSTED_SUSTAINABILITY
Specifications
Description Rotor F-35-8-24
Type Concentrator Plus Rotor
Capacity 8 x 25 mL
Max. Tube Length 90 mm
Max. Tube Diameter 24 mm
For Use With (Equipment) Concentrator Plus Centrifuge