Learn More
Eppendorf™ Concentrator Plus
Shop All Eppendorf Products
Click to view available options
Description:
Basic Concentrator Plus
Concentrator Plus
Concentrator Plus Complete
Concentrator Plus w/ Pump
Electrical Requirements:
230 V, 50/60 Hz (EU)
Includes:
Rotor F-45-48-11
Without Rotor
Unit Size:
Each
Description
Specifications
Specifications
| Frequency | 50/60 Hz |
| Holds | 144 x 0.5 mL Tubes, 96 x 1.5/2 mL, 6 x 50 mL Conical Tubes, 2 Microplate/PCR Plates |
| Includes | Without Rotor |
| Max. Vacuum | 20 hPa (20 Mbar) |
| Pump Type | Integrated Diaphragm Vacuum Pump |
| Type | Concentrators |
| Voltage | 230 V |
| Description | Concentrator Plus w/ Pump |
| Electrical Requirements | 230 V, 50/60 Hz (EU) |
| Speed | 1400 rpm (max. RCF 250 x g) |
| Show More |