Learn More
Novus Biologicals™ Rat Complement Component C5a ELISA Kit (Colorimetric)
Kit (Colorimetric)
Brand:  Novus Biologicals™ NBP3-18863
Product Code. 18672161
  • 7295.00 NOK / 1 set
Specifications
15.63 to 1000 ng/mL
ELISA Kit (Colorimetric)
ELISA, Sandwich ELISA
1 Kit
C5, C5A, HE
9.38 ng/mL
Serum, Plasma, Biological Fluids
727
Storage of components varies. See protocol for specific instructions.
Rat