Learn More
Novus Biologicals™ Rat Complement Component C5a ELISA Kit (Colorimetric)
Kit (Colorimetric)
Brand: Novus Biologicals™ NBP3-18863
This item is not returnable.
View return policy
Description
Specifications
15.63 to 1000 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA, Sandwich ELISA | |
1 Kit | |
C5, C5A, HE |
9.38 ng/mL | |
Serum, Plasma, Biological Fluids | |
727 | |
Storage of components varies. See protocol for specific instructions. | |
Rat |