missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Upright Freezers

Increases flexibility in the laboratory. Tenak™ Comfort Racks for Upright Freezers insure a more ergonomical operating process when working with samples.

Brand:  Tenak™ TE24154

Additional Details : Weight : 5.60000kg

 View more versions of this product

Product Code. 15280724

  • 2360.00 NOK / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Cryoboxes placed in drawers
  • Reduced operating weight
  • Racks can be used with all freezer models
  • Manufactured from stainless steel
  • Sold without cryoboxes
Specifications

Specifications

Comfort Rack
Boxes (35)
Upright Freezers
683 x 140 x 252mm
252 mm
683 mm
7 x 5
Stainless Steel
1 Pack
140 mm
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers