Learn More
Tenak™ Comfort Rack for Upright Freezers
Increases flexibility in the laboratory. Tenak™ Comfort Racks for Upright Freezers insure a more ergonomical operating process when working with samples.
2140.00 NOK - 2520.00 NOK
Specifications
Product Type | Comfort Rack |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Upright Freezers |
No. per Pack | 1 Pack |
Width (Metric) | 140 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15230724
|
Tenak™
TE24136 |
Boxes (32) |
2330.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15240724
|
Tenak™
TE24138 |
Boxes (36) |
2465.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15280724
|
Tenak™
TE24154 |
Boxes (35) |
2360.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15290714
|
Tenak™
TE24126 |
Boxes (24) |
2140.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
15290724
|
Tenak™
TE24156 |
Boxes (40) |
2520.00 NOK
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
Description
- Cryoboxes placed in drawers
- Reduced operating weight
- Racks can be used with all freezer models
- Manufactured from stainless steel
- Sold without cryoboxes
Specifications
Comfort Rack | |
Upright Freezers | |
140 mm |
Stainless Steel | |
1 Pack | |
32 mm |