missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Upright Freezers

Increases flexibility in the laboratory. Tenak™ Comfort Racks for Upright Freezers insure a more ergonomical operating process when working with samples.

Brand:  Tenak™ TE24138

Additional Details : Weight : 5.50000kg

 View more versions of this product

Product Code. 15240724

  • 2465.00 NOK / Each
Estimated Shipment: 22-07-2024
to see stock.
Add to basket

Description

Description

  • Cryoboxes placed in drawers
  • Reduced operating weight
  • Racks can be used with all freezer models
  • Manufactured from stainless steel
  • Sold without cryoboxes
Specifications

Specifications

Comfort Rack
Boxes (36)
Upright Freezers
562 x 140 x 324mm
324 mm
562 mm
9 x 4
Stainless Steel
1 Pack
140 mm
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers