Learn More
Isofield™ Comet Sterile Cleanroom Gloves - Grade A/B Compatible
Description
- Sterile polychloroprene (neoprene) cleanroom gloves
- 300 mm (12 in.) wrist length
- Hand-specific
- Natural color
- Textured fingers and palm
- Beaded cuff
- Gamma irradiation, minimum 25 kGy
- Sterility Assurance Level 10-6
- Latex free for users with latex sensitivities
- Designed for double donning; ideal with Isofield Volta as an under glove
- IPA resistant ink pouches
- Acceptable Quality Level (AQL) 0.65
- Low Particle Count ≥ 0.5 μm (counts/cm2) <1200
- Type B Chemical Splash Protection
Technical and Documentation Compliance:
- COA, COC, COI available for download online
- Chemical innocuousness EN ISO 21420:2020
- Sizing and dexterity EN ISO 21420:2020 and EN ISO 374-2:2019
- Air leak and water leak EN ISO 374-2:2019
- Chemical degradation EN ISO 374-4:2019
- Chemical permeation EN 16523-1:2015+A1:2018
- Viral penetration EN 16604:2004
- Permeation against chemotherapy drugs ASTM D6978-05 (2019)
- Sterility validation test EN ISO 11137 Part 2:2015
- EU type certificate EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN ISO 374-4:2019
- UKCA type certificate EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN ISO 374-4:2019
Specifications
Specifications
| Certifications/Compliance | EN ISO 21420:2020, EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN 455-2:2015 |
| Certifications/Compliance (Europe) | CE Marked; PPE Regulation (EU) 2016/425; Medical Device Regulation (EU) 2017/745 |
| Chemotherapeutic Rated | Yes |
| Chlorinated | Yes |
| Cleanroom Class | Class 100 (ISO 5) |
| Accelerator Free | No |
| AQL | 0.65 |
| Color | Natural |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Show More |