Learn More
Isofield™ Comet Sterile Cleanroom Gloves - Grade A/B Compatible
Product Code. 32016131
Change view
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
32016131 6
32016132 6.5
32016133 7
32016134 7.5
32016135 8
32016136 8.5
32016137 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 32016131 Supplier Isofield™ Supplier No. 3013060
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Latex free and hand specific, the Comet combines flexibility, strength and comfort, and is suitable for double donning as an under glove.
  • Sterile polychloroprene (neoprene) cleanroom gloves
  • 300 mm (12 in.) wrist length
  • Hand-specific
  • Natural color
  • Textured fingers and palm
  • Beaded cuff
  • Gamma irradiation, minimum 25 kGy
  • Sterility Assurance Level 10-6
  • Latex free for users with latex sensitivities
  • Designed for double donning; ideal with Isofield Volta as an under glove
  • IPA resistant ink pouches
  • Acceptable Quality Level (AQL) 0.65
  • Low Particle Count ≥ 0.5 μm (counts/cm2) <1200
  • Type B Chemical Splash Protection
Technical and Documentation Compliance:
  • COA, COC, COI available for download online
  • Chemical innocuousness EN ISO 21420:2020
  • Sizing and dexterity EN ISO 21420:2020 and EN ISO 374-2:2019
  • Air leak and water leak EN ISO 374-2:2019
  • Chemical degradation EN ISO 374-4:2019
  • Chemical permeation EN 16523-1:2015+A1:2018
  • Viral penetration EN 16604:2004
  • Permeation against chemotherapy drugs ASTM D6978-05 (2019)
  • Sterility validation test EN ISO 11137 Part 2:2015
  • EU type certificate EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN ISO 374-4:2019
  • UKCA type certificate EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN ISO 374-4:2019
Specifications
Certifications/Compliance EN ISO 21420:2020, EN ISO 374-1:2016+A1:2018 Type B, EN ISO 374-5:2016, EN 455-2:2015
Certifications/Compliance (Europe) CE Marked; PPE Regulation (EU) 2016/425; Medical Device Regulation (EU) 2017/745
Chemotherapeutic Rated Yes
Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 0.65
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 4.33 mil
Finger Thickness (Metric) 0.14 mm
Hand Specific
Latex Free Yes
Material Polychloroprene
Palm Thickness (English) 5.51 mil
Palm Thickness (Metric) 0.11 mm
Powdered Powder Free
Product Type Cleanroom Gloves
Size 6
Sterility Sterile
Thickness (English) 3.54 mil
Thickness (Metric) 0.09 mm
Grip Design Textured
Manufacturing/QMS Audit Standards ISO 9001
Model Comet
Packaging Type Individually Packed
Length (English) 12 in.
Length (Metric) 300 mm