Learn More
Fisherbrand™ Combs for OmniPAGE Mini Vertical Gel Electrophoresis Unit, 1mm Thick
Shop All Fisherbrand ProductsSpecifications
Specifications
| For Use With (Application) | Mini SDS PAGE, Native PAGE, Gradient, Second Dimension and Nucleic Acid Separations |
| For Use With (Equipment) | OmniPAGE Mini Vertical Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 10 |
| Quantity | 1 Each |
| Thickness (Metric) | 1 mm |
| Volume (Metric) Well | 40 μL |