Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
Product Code. 11873373 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
18
2 Prep, 2 Marker
20
28
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
20 μL
23 μL
25 μL
43 μL
54 μL
59 μL
81 μL
90 μL
243 μL
450 μL
990 μL
Unit Size:
Each
Product Code. 11873373
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
Color Blue
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 12
Quantity 1 Each
Thickness (Metric) 2 mm
Volume (Metric) Sample 81 μL