Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1.5mm Thick
Product Code. 11803383
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
14
16
20
28
30
35
4 Prep, 2 Marker
Volume (Metric) Sample:
11 μL
15 μL
17 μL
19 μL
32 μL
41 μL
44 μL
61 μL
68 μL
182 μL
743 μL
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Volume (Metric) Sample Unit Size
11803383 14 44 μL Each
11853363 10 68 μL Each
11893363 10 44 μL Each
11823373 1 Prep, 1 Marker 743 μL Each
11863373 12 61 μL Each
11843383 16 41 μL Each
11833393 20 32 μL Each
11893393 28 11 μL Each
11803403 28 17 μL Each
11843403 30 19 μL Each
11883403 35 15 μL Each
11823413 4 Prep, 2 Marker 182 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 11803383
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
Color Red
For Use With (Equipment) MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit
Type Comb
No. of Wells 14
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 44 μL