Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1.5mm Thick
Product Code. p-8272012 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
20
25
4 Prep, 2 Marker
8
Volume (Metric) Sample:
15 μL
20 μL
24 μL
34 μL
41 μL
61 μL
115 μL
236 μL
540 μL
Unit Size:
Each
Product Code. 11813313
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Color Red
For Use With (Equipment) Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit
Type Comb
No. of Wells 10
Quantity 1 Each
Thickness (Metric) 1.5 mm
Volume (Metric) Sample 41 μL