Learn More
Fisherbrand™ Combs for Horizontal Tanks MultiSUB Midi Gel Electrophoresis Unit, 1.5mm Thick
Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
12
16
2 Prep, 2 Marker
20
25
4 Prep, 2 Marker
8
Volume (Metric) Sample:
15 μL
20 μL
24 μL
34 μL
41 μL
61 μL
115 μL
236 μL
540 μL
Unit Size:
Each
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10-sample and 20-sample combs are Multichannel pipettor compatible
Specifications
Specifications
| For Use With (Equipment) | Sub-Gel Midi Horizontal Gel Electrophoresis Unit |
| Type | Comb |
| No. of Wells | 4 Prep, 2 Marker |
| Quantity | 1 Each |
| Thickness (Metric) | 1.5 mm |
| Volume (Metric) Sample | 115 μL |