Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 0.75mm Thick
Product Code. 11873443 Shop All Fisherbrand Products
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
16
2 Prep, 2 Marker
20
25
30
36
4 Prep, 2 Marker
40
50
Volume (Metric) Sample:
8 μL
11 μL
13 μL
16 μL
20 μL
30 μL
54 μL
115 μL
236 μL
506 μL
Unit Size:
Each
Product Code. 11873443
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Color Black
For Use With (Equipment) MultiSUB Maxi-Gel Electrophoresis Unit
Type Comb
No. of Wells 30
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Sample 13 μL