Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 0.75mm Thick
Product Code. 11813453 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
16
2 Prep, 2 Marker
20
25
30
36
4 Prep, 2 Marker
40
50
Volume (Metric) Sample:
8 μL
11 μL
13 μL
16 μL
20 μL
30 μL
54 μL
115 μL
236 μL
506 μL
Unit Size:
Each
Product Code. No. of Wells Volume (Metric) Sample unitSize
11813453 36 11 μL Each
11853453 4 Prep, 2 Marker 115 μL Each
11873443 30 13 μL Each
11853433 2 Prep, 2 Marker 236 μL Each
11813433 16 30 μL Each
11833423 1 Prep, 1 Marker 506 μL Each
11843423 10 54 μL Each
11863453 40 8 μL Each
11833463 50 8 μL Each
11863433 20 20 μL Each
11833443 25 16 μL Each
11 options
Product Code. 11813453
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Color Black
For Use With (Equipment) MultiSUB Maxi-Gel Electrophoresis Unit
Type Comb
No. of Wells 36
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Sample 11 μL