Learn More
Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 0.75mm Thick
Product Code. 11813433 Shop All Fisherbrand Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Marker
10
16
2 Prep, 2 Marker
20
25
30
36
4 Prep, 2 Marker
40
50
Volume (Metric) Sample:
8 μL
11 μL
13 μL
16 μL
20 μL
30 μL
54 μL
115 μL
236 μL
506 μL
Unit Size:
Each
Product Code. No. of Wells Volume (Metric) Sample unitSize
11813433 16 30 μL Each
11813453 36 11 μL Each
11853453 4 Prep, 2 Marker 115 μL Each
11873443 30 13 μL Each
11853433 2 Prep, 2 Marker 236 μL Each
11833423 1 Prep, 1 Marker 506 μL Each
11843423 10 54 μL Each
11863453 40 8 μL Each
11833463 50 8 μL Each
11863433 20 20 μL Each
11833443 25 16 μL Each
11 options
Product Code. 11813433
Brand: Fisherbrand™ MS20160.75
Only null left
Add to basket
Add to basket
For use in preparatory electrophoresis and high-throughput screening.
  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications
Color Black
For Use With (Equipment) MultiSUB Maxi-Gel Electrophoresis Unit
Type Comb
No. of Wells 16
Quantity 1 Each
Thickness (Metric) 0.75 mm
Volume (Metric) Sample 30 μL