Learn More
Fisherbrand™ Comb for Wide Format Mini-Plus Horizontal Electrophoresis System
Shop All Fisherbrand ProductsSpecifications
Specifications
| For Use With (Equipment) | Wide Format Mini-Plus Horizontal Gel Unit |
| Type | Comb |
| No. of Wells | 20 |
| Thickness (English) | 0.06 in. |
| Thickness (Metric) | 1.5 mm |
| Volume (Metric) Sample | 32 μL |