Learn More
Nickel Electro™ Clifton™ Stainless Steel Basket
Click to view available options
Capacity (Metric):
1 L
3 L
4 L
6 L
8 L
12 L
14 L
22 L
Dimensions (L x W x H):
177 x 73 x 30 mm
198 x 106 x 50 mm
241 x 260 x 95 mm
250 x 115 x 75 mm
212 x 185 x 110 mm
228 x 107 x 110 mm
250 x 190 x 115 mm
410 x 246 x 110 mm
455 x 250 x 115 mm
Unit Size:
Each
Description
- High quality
- Durable
- Stainless steel
- For use with Ultrasonic Bath
- Includes side lifting handles
Specifications
Specifications
| Type | Basket |
| Capacity (Metric) | 6 L |
| Depth (Metric) | 115 mm |
| For Use With (Application) | For Ultrasonic Bath |
| Height (Metric) | 75 mm |
| Material | Stainless Steel |
| Width (Metric) | 250 mm |
| Dimensions (L x W x H) | 250 x 115 x 75 mm |