Learn More
Nickel Electro™ Clifton™ Stainless Steel Basket
Description
- High quality
- Durable
- Stainless steel
- For use with Ultrasonic Bath
- Includes side lifting handles
Specifications
Specifications
| Type | Basket |
| Capacity (Metric) | 22 L |
| Depth (Metric) | 246 mm |
| For Use With (Application) | For Ultrasonic Bath |
| Height (Metric) | 110 mm |
| Material | Stainless Steel |
| Weight (Metric) | 1.5 kg |
| Width (Metric) | 410 mm |
| Dimensions (L x W x H) | 410 x 246 x 110 mm |