Learn More
Nickel Electro™ Clifton™ Stainless Steel Basket
Description
- High quality
- Durable
- Stainless steel
- For use with Ultrasonic Bath
- Includes side lifting handles
Specifications
Specifications
| Type | Basket |
| Capacity (Metric) | 3 L |
| Depth (Metric) | 106 mm |
| For Use With (Application) | For Ultrasonic Bath |
| Height (Metric) | 50 mm |
| Material | Stainless Steel |
| Width (Metric) | 198 mm |
| Dimensions (L x W x H) | 198 x 106 x 50 mm |