Learn More
Fisherbrand™ Clifton™ Rotary Mixer Discs
Product Code. 12397488 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With: Tubes having Diameters between 25 to 30mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12397488 For Use With: Tubes having Diameters between 25 to 30mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12397488
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get optimum mixing results with Clifton™ tube rotators. Available as fixed or variable speed, the Clifton™ tube rotators are fully adjustable and suitable for continuous or intermittant mixing.
  •  Available as fixed or variable speed models with a broad range of optional accessory heads 
  • Available for mixing micro tubes, tubes, or flasks
  • The range of accessories can also be autoclaved 
  • Suitable for continuous mixing, at variable speeds
  • A small footprint rotator for general use
  • Angle of the mixing disc is fully adjustable from horizontal to vertical
  • Unit is benchtop or wall mountable
Specifications
Product Type Rotary Mixer Disc
For Use With (Equipment) Tubes Having Diameters Between 25 to 30 mm