Learn More
Fisherbrand™ Clear Graduated Glass Bottles with White Polypropylene Cap
691.24 NOK valid until 2024-12-31
Use promo code "21976" to get your promotional price.
Use promo code "21976" to get your promotional price.
Click to view available options
Capacity (English):
1 oz.
2 oz.
4 oz.
8 oz.
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
Description
- Graduated in both milliliters and ounces
- Clear glass bottles are designed with a slight shoulder for easy pouring and handling
- Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Closure Liner | PTFE/Polyethylene Foam |
Closure Material | Polypropylene |
Closure Size | 33-400 mm |
Diameter (Metric) | 34 mm |
Height (Metric) | 68 mm |
Product Type | Graduated Bottle |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Clear |
Diameter (Metric) Outer Neck | 29 mm |