Learn More
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps
GREENER_CHOICE
Product Code. p-7144769 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
Closure Liner:
Polyethylene Cone
Pulp/Tin Foil
Rubber
Unit Size:
Pack of 24
Pack of 288
Pack of 48
Product Code. 16124387
Only null left
Add to basket
Add to basket
Clear USP Type III soda-lime glass with wide mouth for easy removal of contents with graduations in mL and ounces
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (Metric) Outer Neck 29 mm
Diameter (Metric) Inner Neck 20 mm
Height (Metric) With Closure 71 mm
Material Glass (Type 3)
Quantity 48
Closure Color Black
Neck Finish 33-400
Closure Included Yes
Mouth Wide
Height (Metric) 68 mm
Product Type Graduated Bottle
Closure Liner Polyethylene Cone
Closure Material Phenolic
Closure Size 33-400 mm
Diameter (Metric) 34 mm