Learn More
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps
GREENER_CHOICE
Product Code. 16134387 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
60 mL
240 mL
Unit Size:
Pack of 48
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16134387 60 mL Pack of 48
16680654 240 mL Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16134387
Add to basket
Add to basket
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps feature a wide mouth for easy removal of contents.
  • Graduated in both milliliters and ounces
  • Clear glass bottles are designed with a slight shoulder for easy pouring and handling
  • Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Material Glass (Type 3)
Quantity 48
Closure Color Black
Neck Finish 38-400
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Liner Polyethylene Cone
Closure Material Phenolic