Learn More
Fisherbrand™ Clear Glass Wide Mouth Testing Jars with White Polypropylene Caps
GREENER_CHOICE
Product Code. p-7144781 Shop All Fisherbrand Products
Click to view available options
Closure Liner:
PTFE/Polyethylene Foam
Pulp/Vinyl
Unit Size:
Pack of 180
Product Code. 16144547
Only null left
Add to basket
Add to basket
Manufactured from clear soda lime glass wide mouth bottles are ideal for dry and viscous samples
Designed without shoulders for maximum storage capacity
Specifications
Capacity (Metric) 60 mL
Capacity (English) 2.5 oz.
Color Clear
Diameter (Metric) Outer Neck 39 mm
Diameter (Metric) Inner Neck 34 mm
For Use With (Application) Dry and Viscous Samples
Includes Cap
Height (Metric) With Closure 78 mm
Material Glass
Quantity 180
Closure Color White
Neck Finish 43-400
Clarity Clear
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 75 mm
Product Type Testing Jar
Closure Liner PTFE/Polyethylene Foam
Closure Material Polypropylene
Closure Size 43-400 mm
Diameter (Metric) 44 mm
Closure Type Cap