Learn More
Fisherbrand™ Clear French Square Glass Bottles with White Polypropylene Caps
GREENER_CHOICE
Product Code. p-7144766 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
240 mL
480 mL
Closure Liner:
PTFE/Polyethylene Foam
Pulp/Vinyl
Quantity:
24
40
48
84
120
240
576
Unit Size:
Pack of 120
Pack of 24
Pack of 240
Pack of 40
Pack of 48
Pack of 576
Pack of 84
Product Code. Capacity (Metric) Closure Liner Quantity unitSize
16144337 15 mL PTFE/Polyethylene Foam 48 Pack of 48
16174647 15 mL Pulp/Vinyl 576 Pack of 576
16104337 30 mL Pulp/Vinyl 48 Pack of 48
16154337 30 mL PTFE/Polyethylene Foam 48 Pack of 48
16144347 60 mL Pulp/Vinyl 240 Pack of 240
16114337 60 mL Pulp/Vinyl 48 Pack of 48
16164337 60 mL PTFE/Polyethylene Foam 48 Pack of 48
16124337 120 mL Pulp/Vinyl 24 Pack of 24
16154347 120 mL Pulp/Vinyl 120 Pack of 120
16174337 120 mL PTFE/Polyethylene Foam 24 Pack of 24
16134337 240 mL Pulp/Vinyl 24 Pack of 24
16164347 240 mL Pulp/Vinyl 84 Pack of 84
16184337 240 mL PTFE/Polyethylene Foam 24 Pack of 24
16194337 480 mL PTFE/Polyethylene Foam 24 Pack of 24
16174347 480 mL Pulp/Vinyl 40 Pack of 40
15 options
Product Code. 16144337
Only null left
Add to basket
Add to basket
Clear USP Type III soda-lime glass with white polypropylene screw cap ideal for solvent or chemical, sample storage and sample observation
  • Space-saving
  • Easy to fill
  • Clear glass provides maximum content visibility and minimal chemical inference with bottle’s contents
  • Square design allows the bottle to hold the maximum volume of contents while taking up minimum shelf and storage space
  • Wide mouth opening is ideal for mixing, storing and sampling
Specifications
Capacity (Metric) 15 mL
Capacity (English) 0.5 oz.
Color Clear
Diameter (Metric) Outer Neck 18 mm
Diameter (Metric) Inner Neck 10 mm
Height (Metric) With Closure 65 mm
Material Glass (Type 3)
Quantity 48
Closure Color White
Neck Finish 20-400
Closure Included Yes
Mouth Wide
Height (Metric) 62 mm
Product Type French Square Bottle
Closure Liner PTFE/Polyethylene Foam
Closure Material Polypropylene
Closure Size 20-400 mm
Diameter (Metric) 26 mm