Learn More
Fisherbrand™ Clear Boston Round Glass Bottles without Caps
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
950 mL
Unit Size:
Pack of 108
Pack of 12
Pack of 160
Pack of 288
Pack of 432
Pack of 60
Description
- General use bottles ideal for when content integrity and sample or product visibility is important
- Narrow screw neck opening, curved shoulders, and a round body
- Clear glass offers minimal chemical interference with contents and is appropriate when visibility is important but protection from UV rays is not necessary
- Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Clear |
| Diameter (Metric) Outer Neck | 18 mm |
| Diameter (Metric) Inner Neck | 10 mm |
| Material | Glass (Type 3) |
| Quantity | 288 |
| Neck Finish | 20-400 |
| Closure Included | No |
| Mouth | Narrow |
| Show More |