Learn More
Sartorius Claristep™ Single Tray
Shop All Sartorius Products
Click to view available options
:
Claristep™ Single Tray; Capacity: 600 μL
Description
Includes
Single Tray only
- Multiplex up to 8 samples
- No vacuum source needed
- No power supply needed
- Yellow
Specifications
Specifications
| Capacity (Metric) | 600 μL |
| Kit Contents | Single Tray Only |
| Content And Storage | Room Temperature |
| Quantity | 1 Ea. |
| For Use With (Application) | HPLC |
| Type | Tools and Accessories |
| Sufficient For | Analytical Sample Preparation, Clarification |
| Format | 8-well Strips |