Learn More
R&L Enterprises™ Semi-Micro Burette Clamp for Stands
For ease of connection on ring stands. R&L Enterprises™ Semi-Micro Burette Clamp for Stands comprises two 13mm plastic-coated spring clips mounted 50mm apart on a plate attached to a rod.
Brand:  R&L Enterprises™ 3595
Additional Details : Weight : 1.00000kg
Product Code. 12996826
  • 122.00 NOK / Each
Estimated Shipment: 12-06-2024
Description
  • To fit bossheads
  • For 25 or 50mL burettes
  • Rod is 150 to 8mm diameter
Specifications
Specifications
1 Ea.
Clamp
For use with 25/50mL burettes