missing translation for 'onlineSavingsMsg'
Learn More

CISD2 Mouse anti-Human, Mouse, Rat, Zebrafish, Clone: 3D7A3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66082-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847635

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This monoclonal antibody is specific to CISD2 and does not cross-react with CISD1.

Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype.
Specifications

Specifications

CISD2
Monoclonal
1.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7T326, Q8N5K1, Q9CQB5
cisd2
Protein A
RUO
295457, 393354, 493856, 67006
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
3D7A3
Unconjugated
CISD2
CDGSH iron sulfur domain 2, CDGSH2, CISD2, CISD2-Specific, ERIS, Miner1, MitoNEET related 1 protein, NAF 1, WFS2, ZCD2
Mouse
150 μL
Primary
Human, Mouse, Rat, Zebrafish
Antibody
IgG2b
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers