missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD2 Mouse anti-Human, Mouse, Rat, Zebrafish, Clone: 3D7A3, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | CISD2 |
|---|---|
| Clone | 3D7A3 |
| Concentration | 1.65 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857635
|
Proteintech
66082-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847635
|
Proteintech
66082-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This monoclonal antibody is specific to CISD2 and does not cross-react with CISD1.
Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype.Specifications
| CISD2 | |
| 1.65 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CDGSH iron sulfur domain 2, CDGSH2, CISD2, CISD2-Specific, ERIS, Miner1, MitoNEET related 1 protein, NAF 1, WFS2, ZCD2 | |
| cisd2 | |
| Primary | |
| -20°C | |
| CISD2 |
| 3D7A3 | |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Zebrafish, Rat | |
| Q7T326, Q8N5K1, Q9CQB5 | |
| 295457, 393354, 493856, 67006 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title