missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12013-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.Specifications
CHST10 | |
Polyclonal | |
Unconjugated | |
CHST10 | |
CHST10, HNK 1 sulfotransferase, HNK 1ST, HNK1ST, HuHNK 1ST | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140568, 9486, 98388 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43529, O54702, Q6PGK7 | |
CHST10 | |
CHST10 Fusion Protein Ag2627 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |