missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | CHST10 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899463
|
Proteintech
12013-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16889463
|
Proteintech
12013-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.Specifications
CHST10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43529, O54702, Q6PGK7 | |
140568, 9486, 98388 | |
CHST10 Fusion Protein Ag2627 | |
Primary | |
-20°C | |
CHST10 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CHST10, HNK 1 sulfotransferase, HNK 1ST, HNK1ST, HuHNK 1ST | |
CHST10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |