Learn More
Novus Biologicals Human DC-SIGN/CD209 ELISA Kit (Colorimetric)
Product Code. 18609770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18609770 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18609770 Supplier Novus Biologicals Supplier No. NBP280334
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Assay Range 31.25-2000 pg/mL
Assay Sensitivity 11.8 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Human DC-SIGN/CD209.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 30835
Gene Symbol CD209
Quantity 1 Kit
Research Discipline Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Storage Requirements Storage is content dependent.
Synonym CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Target Human