Learn More
Chromium(III) Acetylacetonate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 21679-31-2 |
| Melting Point | 213°C to 216°C |
| MDL Number | MFCD00000015 |
| Quantity | 100 g |
| Synonym | Chromium(III) acetylacetonate, Tris(acetylacetonato)chromium(III), Tris(2,4-pentanedionato)chromium(III), Chromium(III) acetylacetonate |
| Molecular Weight (g/mol) | 349.32 |
| Grade | Research |