missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | CHRNA9 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16844555
|
Proteintech
26025-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16834555
|
Proteintech
26025-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHRNA9 is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor superfamily. CHRNA9 is a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. It is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea.This gene is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor gene superfamily. It encodes a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. This protein is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea. The protein is additionally expressed in keratinocytes, the pituitary gland, B-cells and T-cells.Specifications
| CHRNA9 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q9UGM1 | |
| 231252, 55584 | |
| CHRNA9 Fusion Protein Ag23272 | |
| Primary | |
| -20°C | |
| CHRNA9 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CHRNA9, HSA243342, NACHR alpha 9, NACHRA9 | |
| CHRNA9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title