Learn More
CHRNA9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26025-1-AP-150UL
This item is not returnable.
View return policy
Description
CHRNA9 is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor superfamily. CHRNA9 is a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. It is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea.This gene is a member of the ligand-gated ionic channel family and nicotinic acetylcholine receptor gene superfamily. It encodes a plasma membrane protein that forms homo- or hetero-oligomeric divalent cation channels. This protein is involved in cochlea hair cell development and is also expressed in the outer hair cells (OHCs) of the adult cochlea. The protein is additionally expressed in keratinocytes, the pituitary gland, B-cells and T-cells.Specifications
| CHRNA9 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA9 | |
| CHRNA9, HSA243342, NACHR alpha 9, NACHRA9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231252, 55584 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UGM1 | |
| CHRNA9 | |
| CHRNA9 Fusion Protein Ag23272 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |